2-Methyl-5-pyridineboronic acid pinacol ester ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Methylpyridine-5-boronic acid pinacol ester, 6-Methylpyridine-3-boronic acid pinacol ester, 2-(6-Methyl-3-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 6-Methyl-3-pyridylboronic acid Pinacol ester

069078-1G 069078-5G
101938-830EA 186.15 USD
101938-830 101938-832
2-Methyl-5-pyridineboronic acid pinacol ester ≥97%
2-Methyl-5-pyridineboronic acid pinacol ester

Hazard: Irritant

Formula: C₁₂H₁₈BNO₂
MDL Number: MFCD08669608
CAS Number: 610768-32-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR