Copper(I) 2-hydroxy-3-methylbenzoate ≥96%

Supplier: Matrix Scientific

Synonyms: Copper(I)-3-methylsalicylate

069077-5G 069077-25G
101938-826EA 184.62 USD
101938-826 101938-828
Copper(I) 2-hydroxy-3-methylbenzoate ≥96%
Copper(I) 2-hydroxy-3-methylbenzoate

Hazard: Irritant

Formula: C₈H₇CuO₃
MW: 214.69 g/mol
MDL Number: MFCD10699180
CAS Number: 326477-70-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR