trans-3,3-Dimethyl-1-butenylboronic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 2-tert-Butyl-trans-vinylboronic acid, (E)-(3,3-Dimethylbut-1-en-1-yl)boronic acid

069073-1G 069073-5G 069073-25G
101938-802EA 283.08 USD
101938-802 101938-804 101938-806
trans-3,3-Dimethyl-1-butenylboronic acid ≥97%
trans-3,3-Dimethyl-1-butenylboronic acid

Hazard: Irritant

Formula: C₆H₁₃BO₂
MW: 127.98 g/mol
MDL Number: MFCD01074624
CAS Number: 86595-37-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR