Thiazole-2-carbaldehyde ≥98%
Supplier: Matrix Scientific
Synonyms:
2-Thiazolecarboxaldehyde, Thiazole-2-carboxaldehyde
069072-1G
069072-5G
069072-25G
101938-796EA
15.38
USD
101938-796
101938-798
101938-800
Thiazole-2-carbaldehyde ≥98%
Thiazole-2-carbaldehyde
Hazard: Irritant
Formula:
C₄H₃NOS MW: 113.13 g/mol |
MDL Number:
MFCD00142924 CAS Number: 10200-59-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...