3-Isopropoxy-4-(tributylstannyl)-1,2-cyclobutenedione ≥97%

Supplier: Matrix Scientific

Synonyms: 3-Isopropoxy-4-(tri-n-butylstannyl)-3-cyclobutene-1,2-dione

069071-250MG 069071-500MG 069071-1G
101938-790EA 776.92 USD
101938-790 101938-792 101938-794
3-Isopropoxy-4-(tributylstannyl)-1,2-cyclobutenedione ≥97%
3-Isopropoxy-4-(tributylstannyl)-1,2-cyclobutenedione

Hazard: Irritant

Formula: C₁₉H₃₄O₃Sn
MDL Number: MFCD01319011
CAS Number: 129034-70-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR