5-((6-Chloropyridazin-3-yl)methyl)-2-(2-fluorophenyl)-5H-imidazo[4,5-c]pyridine ≥95%
Supplier: Matrix Scientific
069059-250MG
069059-500MG
101938-768EA
669.23
USD
101938-768
101938-770
5-((6-Chloropyridazin-3-yl)methyl)-2-(2-fluorophenyl)-5H-imidazo[4,5-c]pyridine ≥95%
5-((6-Chloropyridazin-3-yl)methyl)-2-(2-fluorophenyl)-5H-imidazo[4,5-c]pyridine
Hazard: Irritant
Formula:
C₁₇H₁₁ClFN₅ |
MDL Number:
MFCD19443922 CAS Number: 1000787-76-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...