N-(6-Bromonaphthalen-2-yl)methanesulfonamide ≥95%

Supplier: Matrix Scientific

069053-250MG 069053-500MG
101938-752EA 669.23 USD
101938-752 101938-754
N-(6-Bromonaphthalen-2-yl)methanesulfonamide ≥95%
N-(6-Bromonaphthalen-2-yl)methanesulfonamide

Hazard: Irritant

Formula: C₁₁H₁₀BrNO₂S
MDL Number: MFCD13191640
CAS Number: 1132940-86-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR