4-(1-Methylpiperidin-4-yl)benzoic acid ≥95%

Supplier: Matrix Scientific

069052-1G 069052-5G
101938-748EA 1169.23 USD
101938-748 101938-750
4-(1-Methylpiperidin-4-yl)benzoic acid ≥95%
4-(1-Methylpiperidin-4-yl)benzoic acid

Hazard: Irritant

Formula: C₁₃H₁₇NO₂
MDL Number: MFCD13191658
CAS Number: 281234-85-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR