tert-Butyl-4-(2,2,2-trifluoroacetamido)piperazine-1-carboxylate ≥95%

Supplier: Matrix Scientific

069044-250MG 069044-500MG
101938-734EA 436.92 USD
101938-734 101938-736
tert-Butyl-4-(2,2,2-trifluoroacetamido)piperazine-1-carboxylate ≥95%
tert-Butyl-4-(2,2,2-trifluoroacetamido)piperazine-1-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₈F₃N₃O₃
MW: 297.28 g/mol
MDL Number: MFCD12912661
CAS Number: 1198286-22-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR