1-(4-(2,4-Difluorobenzoyl)piperidin-1-yl)-2,2,2-trifluoroethanone ≥95%

Supplier: Matrix Scientific

069041-250MG 069041-500MG 069041-1G
101938-722EA 263.08 USD
101938-722 101938-724 101938-726
1-(4-(2,4-Difluorobenzoyl)piperidin-1-yl)-2,2,2-trifluoroethanone ≥95%
1-(4-(2,4-Difluorobenzoyl)piperidin-1-yl)-2,2,2-trifluoroethanone

Hazard: Irritant

Formula: C₁₄H₁₂F₅NO₂
MDL Number: MFCD12912617
CAS Number: 1198287-09-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR