tert-Butyl-2-methyl-3-oxopiperazine-1-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: 1-Boc-2-methyl-3-oxopiperazine

069035-1G 069035-5G
101938-710EA 1512.31 USD
101938-710 101938-712
tert-Butyl-2-methyl-3-oxopiperazine-1-carboxylate ≥95%
tert-Butyl-2-methyl-3-oxopiperazine-1-carboxylate

Hazard: Irritant

Formula: C₁₀H₁₈N₂O₃
MW: 214.27 g/mol
MDL Number: MFCD06803995
CAS Number: 76003-30-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR