(2S)-4-Oxopiperidine-2-carboxylic acid hydrochloride ≥95%

Supplier: Matrix Scientific

069033-250MG 069033-500MG 069033-1G
101938-700EA 1106.15 USD
101938-700 101938-702 101938-704
(2S)-4-Oxopiperidine-2-carboxylic acid hydrochloride ≥95%
(2S)-4-Oxopiperidine-2-carboxylic acid hydrochloride

Hazard: Irritant

Formula: C₆H₁₀ClNO₃
MDL Number: MFCD12912671
CAS Number: 65060-18-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR