2,2,2-Trifluoro-1-(4-(4-bromobenzoyl)piperidin-1-yl)ethanone ≥95%

Supplier: Matrix Scientific

069030-250MG 069030-500MG 069030-1G
101938-688EA 263.08 USD
101938-688 101938-690 101938-692
2,2,2-Trifluoro-1-(4-(4-bromobenzoyl)piperidin-1-yl)ethanone ≥95%
2,2,2-Trifluoro-1-(4-(4-bromobenzoyl)piperidin-1-yl)ethanone

Hazard: Irritant

Formula: C₁₄H₁₃BrF₃NO₂
MDL Number: MFCD12912608
CAS Number: 203186-01-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR