2,2,2-Trifluoro-1-(4-benzoylpiperidin-1-yl)ethanone ≥95%

Supplier: Matrix Scientific

069029-250MG 069029-500MG 069029-1G
101938-682EA 263.08 USD
101938-682 101938-684 101938-686
2,2,2-Trifluoro-1-(4-benzoylpiperidin-1-yl)ethanone ≥95%
2,2,2-Trifluoro-1-(4-benzoylpiperidin-1-yl)ethanone

Hazard: Irritant

Formula: C₁₄H₁₄F₃NO₂
MDL Number: MFCD00203351
CAS Number: 257946-67-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR