tert-Butyl-4-phenyl-3,6-dihydro-2H-pyridine-1-carboxylate ≥95%

Supplier: Matrix Scientific

069025-500MG 069025-1G 069025-5G
101938-668EA 349.23 USD
101938-668 101938-670 101938-672
tert-Butyl-4-phenyl-3,6-dihydro-2H-pyridine-1-carboxylate ≥95%
tert-Butyl-4-phenyl-3,6-dihydro-2H-pyridine-1-carboxylate

Hazard: Irritant

Formula: C₁₆H₂₁NO₂
MW: 259.35 g/mol
MDL Number: MFCD11977149
CAS Number: 186347-72-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR