tert-Butyl-4-hydroxy-4-phenylpiperidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

069024-1G 069024-5G
101938-664EA 378.46 USD
101938-664 101938-666
tert-Butyl-4-hydroxy-4-phenylpiperidine-1-carboxylate ≥95%
tert-Butyl-4-hydroxy-4-phenylpiperidine-1-carboxylate

Hazard: Irritant

Formula: C₁₆H₂₃NO₃
MW: 277.37 g/mol
MDL Number: MFCD09953388
CAS Number: 172734-33-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR