1-tert-Butyl-2-methyl-2-(3-chloropropyl)pyrrolidine-1,2-dicarboxylate ≥95%

Supplier: Matrix Scientific

069022-250MG 069022-500MG
101938-660EA 524.62 USD
101938-660 101938-662
1-tert-Butyl-2-methyl-2-(3-chloropropyl)pyrrolidine-1,2-dicarboxylate ≥95%
1-tert-Butyl-2-methyl-2-(3-chloropropyl)pyrrolidine-1,2-dicarboxylate

Hazard: Irritant

Formula: C₁₄H₂₄ClNO₄
MDL Number: MFCD12912662
CAS Number: 909076-34-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR