Methyl-1-benzylpyrrolidine-3-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 1-benzylpyrrolidine-3-carboxylate

069019-1G 069019-5G 069019-25G
101938-654EA 92.31 USD
101938-654 101938-656 101938-658
Methyl-1-benzylpyrrolidine-3-carboxylate ≥95%
Methyl-1-benzylpyrrolidine-3-carboxylate

Hazard: Irritant

Formula: C₁₃H₁₇NO₂
MW: 219.28 g/mol
MDL Number: MFCD04038678
CAS Number: 17012-21-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR