tert-Butyl-7-benzyl-3,7-diazabicyclo[3.3.1]nonane-3-carboxylate ≥97%

Supplier: Matrix Scientific

069016-250MG 069016-500MG
101938-644EA 1629.23 USD
101938-644 101938-646
tert-Butyl-7-benzyl-3,7-diazabicyclo[3.3.1]nonane-3-carboxylate ≥97%
tert-Butyl-7-benzyl-3,7-diazabicyclo[3.3.1]nonane-3-carboxylate

Hazard: Irritant

Formula: C₁₉H₂₈N₂O₂
MW: 316.45 g/mol
MDL Number: MFCD19443918
CAS Number: 227940-71-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR