Ethyl (1S,4R)-3-benzyl-3-azabicyclo[2.2.1]heptane-7-carboxylate ≥95%

Supplier: Matrix Scientific

069014-250MG 069014-1G
101938-636EA 640 USD
101938-636 101938-638
Ethyl (1S,4R)-3-benzyl-3-azabicyclo[2.2.1]heptane-7-carboxylate ≥95%
Ethyl (1S,4R)-3-benzyl-3-azabicyclo[2.2.1]heptane-7-carboxylate

Hazard: Irritant

Formula: C₁₆H₂₁NO₂
MDL Number: MFCD20486942
CAS Number: 745836-32-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR