tert-Butyl-4-(1H-indol-1-yl)piperidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 4-(1H-indol-1-yl)piperidine-1-carboxylate

069012-250MG 069012-500MG 069012-1G
101938-624EA 698.46 USD
101938-624 101938-626 101938-628
tert-Butyl-4-(1H-indol-1-yl)piperidine-1-carboxylate ≥95%
tert-Butyl-4-(1H-indol-1-yl)piperidine-1-carboxylate

Hazard: Irritant

Formula: C₁₈H₂₄N₂O₂
MW: 300.4 g/mol
MDL Number: MFCD09833914
CAS Number: 170364-89-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR