2-Benzyl-2-azabicyclo[2.2.1]hept-5-ene ≥95%
Supplier: Matrix Scientific
069011-1G
069011-5G
101938-620EA
349.23
USD
101938-620
101938-622
2-Benzyl-2-azabicyclo[2.2.1]hept-5-ene ≥95%
2-Benzyl-2-azabicyclo[2.2.1]hept-5-ene
Hazard: Irritant
Formula:
C₁₃H₁₅N |
MDL Number:
MFCD00180650 CAS Number: 112375-05-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...