1-Benzyl-4-(nitromethyl)piperidin-4-ol
Supplier: Matrix Scientific
068993-500MG
068993-1G
068993-5G
101938-570EA
436.92
USD
101938-570
101938-572
101938-574
1-Benzyl-4-(nitromethyl)piperidin-4-ol
1-Benzyl-4-(nitromethyl)piperidin-4-ol
Hazard: Irritant
Formula:
C₁₃H₁₈N₂O₃ |
MDL Number:
MFCD11840355 CAS Number: 34259-89-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...