1-Benzyl-3-bromopiperidin-4-one hydrobromide ≥95%

Supplier: Matrix Scientific

068988-500MG 068988-1G 068988-5G
101938-558EA 466.15 USD
101938-558 101938-560 101938-562
1-Benzyl-3-bromopiperidin-4-one hydrobromide ≥95%
1-Benzyl-3-bromopiperidin-4-one hydrobromide

Hazard: Irritant

Formula: C₁₂H₁₅Br₂NO
MDL Number: MFCD16040214
CAS Number: 83877-88-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR