4-(Piperidin-3-ylthio)pyridine dihydrochloride ≥95%

Supplier: Matrix Scientific

068984-250MG 068984-500MG 068984-1G
101938-552EA 698.46 USD
101938-552 101938-554 101938-556
4-(Piperidin-3-ylthio)pyridine dihydrochloride ≥95%
4-(Piperidin-3-ylthio)pyridine dihydrochloride

Hazard: Irritant

Formula: C₁₀H₁₆Cl₂N₂S
MDL Number: MFCD10566042
CAS Number: 1198283-96-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR