2-(4-Piperidyl)acetonitrile hydrochloride ≥95%

Supplier: Matrix Scientific

068983-250MG 068983-500MG 068983-1G
101938-546EA 384.62 USD
101938-546 101938-548 101938-550
2-(4-Piperidyl)acetonitrile hydrochloride ≥95%
2-(4-Piperidyl)acetonitrile hydrochloride

Hazard: Irritant

Formula: C₇H₁₃ClN₂
MDL Number: MFCD12026392
CAS Number: 153566-98-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR