Benzyl-3-(tert-butoxycarbonylamino)piperidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

068982-500MG 068982-1G 068982-5G
101938-540EA 901.54 USD
101938-540 101938-542 101938-544
Benzyl-3-(tert-butoxycarbonylamino)piperidine-1-carboxylate ≥95%
Benzyl-3-(tert-butoxycarbonylamino)piperidine-1-carboxylate

Hazard: Irritant

Formula: C₁₈H₂₆N₂O₄
MW: 334.42 g/mol
MDL Number: MFCD09952600
CAS Number: 406213-47-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR