tert-Butyl-4-[acetyl(methyl)amino]piperidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

068981-500MG 068981-1G 068981-5G
101938-534EA 901.54 USD
101938-534 101938-536 101938-538
tert-Butyl-4-[acetyl(methyl)amino]piperidine-1-carboxylate ≥95%
tert-Butyl-4-[acetyl(methyl)amino]piperidine-1-carboxylate

Hazard: Irritant

Formula: C₁₃H₂₄N₂O₃
MW: 256.35 g/mol
MDL Number: MFCD09908272
CAS Number: 197727-57-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR