3,3,5-Tribromo-1H-pyrrolo[2,3-b]pyridin-2-one ≥95%

Supplier: Matrix Scientific

Synonyms: 3,3,5-Tribromo-1H-pyrrolo[2,3-b]pyridin-2(3H)-one

068978-100MG 068978-250MG 068978-500MG
101938-516EA 407.69 USD
101938-516 101938-518 101938-520
3,3,5-Tribromo-1H-pyrrolo[2,3-b]pyridin-2-one ≥95%
3,3,5-Tribromo-1H-pyrrolo[2,3-b]pyridin-2-one

Hazard: Irritant

Formula: C₇H₃Br₃N₂O
MW: 370.84 g/mol
MDL Number: MFCD09028431
CAS Number: 183208-32-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR