1-Benzyl-3-bromopyrrolo[2,3-b]pyridine ≥95%

Supplier: Matrix Scientific

068977-250MG 068977-500MG 068977-1G
101938-510EA 815.38 USD
101938-510 101938-512 101938-514
1-Benzyl-3-bromopyrrolo[2,3-b]pyridine ≥95%
1-Benzyl-3-bromopyrrolo[2,3-b]pyridine

Hazard: Irritant

Formula: C₁₄H₁₁BrN₂
MDL Number: MFCD09834871
CAS Number: 281192-93-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR