5-Pyrimidinecarbonyl chloride ≥95%

Supplier: Matrix Scientific

068973-1G 068973-5G
101938-490EA 183.08 USD
101938-490 101938-492
5-Pyrimidinecarbonyl chloride ≥95%
5-Pyrimidinecarbonyl chloride

Hazard: Irritant

Formula: C₅H₃ClN₂O
MW: 142.55 g/mol
MDL Number: MFCD03789627
CAS Number: 40929-48-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR