1-(1-Benzylpiperidin-4-yl)-2,2,2-trifluoroethanone oxalate ≥95%

Supplier: Matrix Scientific

068968-500MG 068968-1G
101938-464EA 815.38 USD
101938-464 101938-466
1-(1-Benzylpiperidin-4-yl)-2,2,2-trifluoroethanone oxalate ≥95%
1-(1-Benzylpiperidin-4-yl)-2,2,2-trifluoroethanone oxalate

Hazard: Irritant

Formula: C₁₆H₁₈F₃NO₅
MDL Number: MFCD12026432
CAS Number: 1198286-70-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR