tert-Butyl-4-(1-cyanopropylidene)piperidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

068965-500MG 068965-1G
101938-456EA 756.92 USD
101938-456 101938-458
tert-Butyl-4-(1-cyanopropylidene)piperidine-1-carboxylate ≥95%
tert-Butyl-4-(1-cyanopropylidene)piperidine-1-carboxylate

Hazard: Irritant

Formula: C₁₄H₂₂N₂O₂
MDL Number: MFCD12026389
CAS Number: 1198283-75-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR