1'-Benzyl-7-methoxyspiro[chromane-2,4'-piperidine]-4-one ≥95%

Supplier: Matrix Scientific

068960-500MG 068960-1G
101938-448EA 263.08 USD
101938-448 101938-450
1'-Benzyl-7-methoxyspiro[chromane-2,4'-piperidine]-4-one ≥95%
1'-Benzyl-7-methoxyspiro[chromane-2,4'-piperidine]-4-one

Hazard: Irritant

Formula: C₂₁H₂₃NO₃
MDL Number: MFCD08311603
CAS Number: 868361-89-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR