Ethyl-4-hydroxy-5-isopropylpyrrolo[1,2-f][1,2,4]triazine-6-carboxylate ≥95%

Supplier: Matrix Scientific

068932-250MG 068932-1G
101938-320EA 901.54 USD
101938-320 101938-324
Ethyl-4-hydroxy-5-isopropylpyrrolo[1,2-f][1,2,4]triazine-6-carboxylate ≥95%
Ethyl-4-hydroxy-5-isopropylpyrrolo[1,2-f][1,2,4]triazine-6-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₅N₃O₃
MW: 249.27 g/mol
MDL Number: MFCD11976589
CAS Number: 651744-40-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR