3-(Hydroxymethyl)cyclobutanone ≥95%

Supplier: Matrix Scientific

068928-250MG 068928-5G
101938-302EA 621.54 USD
101938-302 101938-306
3-(Hydroxymethyl)cyclobutanone ≥95%
3-(Hydroxymethyl)cyclobutanone

Hazard: Irritant

Formula: C₅H₈O₂
MW: 100.12 g/mol
MDL Number: MFCD11100969
CAS Number: 183616-18-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR