L-Azetidine-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: L-Trimethyleneimine-2-carboxylic Acid, (S)-Azetidine-2-carboxylic acid, (S)-(-)-2-Azetidinecarboxylic acid, L-Azetidine-2-carboxylic Acid [Antagonist of L-Proline]

068916-500MG 068916-5G
101938-240EA 349.23 USD
101938-240 101938-244
L-Azetidine-2-carboxylic acid ≥95%
L-Azetidine-2-carboxylic acid

Hazard: Irritant

Formula: C₄H₇NO₂
MW: 101.11 g/mol
MDL Number: MFCD00005166
CAS Number: 2133-34-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR