Nortropinone hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: 8-Azabicyclo[3.2.1]octan-3-one hydrochloride

068913-1G 068913-25G
101938-224EA 116.92 USD
101938-224 101938-228
Nortropinone hydrochloride ≥95%
Nortropinone hydrochloride

Hazard: Irritant

Formula: C₇H₁₂ClNO
MW: 161.63 g/mol
MDL Number: MFCD03613582
CAS Number: 25602-68-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR