Tetrahydro-2H-pyran-3-carboxylic acid ≥95%

Supplier: Matrix Scientific

068909-1G 068909-5G
101938-208EA 1430.77 USD
101938-208 102551-726
Tetrahydro-2H-pyran-3-carboxylic acid ≥95%
Tetrahydro-2H-pyran-3-carboxylic acid

Hazard: Irritant

Formula: C₆H₁₀O₃
MW: 130.14 g/mol
MDL Number: MFCD04114349
CAS Number: 873397-34-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR