3-Aminomethyl-3-[bis(phenylmethyl)amino]oxetane ≥95%

Supplier: Matrix Scientific

068895-500MG 068895-5G
101938-134EA 407.69 USD
101938-134 101938-138
3-Aminomethyl-3-[bis(phenylmethyl)amino]oxetane ≥95%
3-Aminomethyl-3-[bis(phenylmethyl)amino]oxetane

Hazard: Irritant

Formula: C₁₈H₂₂N₂O
MW: 282.39 g/mol
MDL Number: MFCD19443904
CAS Number: 1021392-84-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR