(E)-3-(2,3,4-Trihydroxyphenyl)prop-2-enoic acid ≥95%

Supplier: Matrix Scientific

068871-500MG 068871-5G
101938-024EA 872.31 USD
101938-024 101938-028
(E)-3-(2,3,4-Trihydroxyphenyl)prop-2-enoic acid ≥95%
(E)-3-(2,3,4-Trihydroxyphenyl)prop-2-enoic acid

Hazard: Irritant

Formula: C₉H₈O₅
MDL Number: MFCD03002815
CAS Number: 13058-13-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR