1-(2-Fluorobenzyl)-1H-pyrazolo[3,4-b]pyridine-3-carboxamidine hydrochloride ≥95%
Supplier: Matrix Scientific
Synonyms:
1-(2-Fluorobenzyl)-1H-pyrazolo[3,4-b]pyridine-3-carboximidamide hydrochloride
068862-250MG
068862-1G
101937-986EA
768.74
USD
101937-986
101937-990
1-(2-Fluorobenzyl)-1H-pyrazolo[3,4-b]pyridine-3-carboxamidine hydrochloride ≥95%
1-(2-Fluorobenzyl)-1H-pyrazolo[3,4-b]pyridine-3-carboxamidine hydrochloride
Hazard: Irritant
Formula:
C₁₄H₁₃ClFN₅ MW: 305.74 g/mol |
MDL Number:
MFCD18432772 CAS Number: 256499-19-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...