2-Methyl-1H-pyrrolo[2,3-b]pyridine ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Methyl-7-azaindole

068846-250MG 068846-1G 068846-5G
101937-916EA 349.23 USD
101937-916 102833-438 102833-440
2-Methyl-1H-pyrrolo[2,3-b]pyridine ≥95%
2-Methyl-1H-pyrrolo[2,3-b]pyridine

Hazard: Irritant

Formula: C₈H₈N₂
MW: 132.1600 g/mol
MDL Number: MFCD08669524
CAS Number: 23612-48-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR