2,4-Dichloro-6-methyl-7H-pyrrolo[2,3-d]pyrimidine ≥95%

Supplier: Matrix Scientific

068798-250MG 068798-5G
101937-734EA 461.54 USD
101937-734 102833-436
2,4-Dichloro-6-methyl-7H-pyrrolo[2,3-d]pyrimidine ≥95%
2,4-Dichloro-6-methyl-7H-pyrrolo[2,3-d]pyrimidine

Hazard: Irritant

Formula: C₇H₅Cl₂N₃
MDL Number: MFCD13193622
CAS Number: 1192711-71-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR