4-Chloro-1H-pyrrolo[2,3-c]pyridine ≥95%
Supplier: Matrix Scientific
068777-250MG
068777-1G
101937-632EA
1092.31
USD
101937-632
101937-636
4-Chloro-1H-pyrrolo[2,3-c]pyridine ≥95%
4-Chloro-1H-pyrrolo[2,3-c]pyridine
Hazard: Irritant
Formula:
C₇H₅ClN₂ MW: 152.58 g/mol |
MDL Number:
MFCD16657181 CAS Number: 1188313-15-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...