Methyl-3-bromo-1H-pyrazolo[3,4-b]pyridine-5-carboxylate ≥95%

Supplier: Matrix Scientific

068756-250MG 068756-1G
101937-542EA 944.62 USD
101937-542 101937-546
Methyl-3-bromo-1H-pyrazolo[3,4-b]pyridine-5-carboxylate ≥95%
Methyl-3-bromo-1H-pyrazolo[3,4-b]pyridine-5-carboxylate

Hazard: Irritant

Formula: C₈H₆BrN₃O₂
MDL Number: MFCD17016018
CAS Number: 1221288-27-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR