4-Chloro-6-iodo-7H-pyrrolo[2,3-d]pyrimidine ≥95%

Supplier: Matrix Scientific

068745-250MG 068745-1G
101937-490EA 844.62 USD
101937-490 101937-494
4-Chloro-6-iodo-7H-pyrrolo[2,3-d]pyrimidine ≥95%
4-Chloro-6-iodo-7H-pyrrolo[2,3-d]pyrimidine

Hazard: Irritant

Formula: C₆H₃ClIN₃
MW: 279.4700 g/mol
MDL Number: MFCD11518919
CAS Number: 876343-10-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR