2-Methylindazole-7-carboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Methyl-2H-indazole-7-carboxylic acid

068728-250MG 068728-1G
101937-416EA 436.92 USD
101937-416 101937-420
2-Methylindazole-7-carboxylic acid ≥95%
2-Methylindazole-7-carboxylic acid

Hazard: Irritant

Formula: C₉H₈N₂O₂
MDL Number: MFCD15071445
CAS Number: 1234615-75-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR