5-Chloro-1H-pyrrolo[2,3-c]pyridin-2(3H)-one ≥95%

Supplier: Matrix Scientific

068717-250MG 068717-1G
101937-362EA 1629.23 USD
101937-362 101937-366
5-Chloro-1H-pyrrolo[2,3-c]pyridin-2(3H)-one ≥95%
5-Chloro-1H-pyrrolo[2,3-c]pyridin-2(3H)-one

Hazard: Irritant

Formula: C₇H₅ClN₂O
MW: 168.5800 g/mol
MDL Number: MFCD11518942
CAS Number: 136888-17-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR