4-Chloro-7H-pyrrolo[2,3-d]pyrimidine-5-carboxylic acid ≥95%

Supplier: Matrix Scientific

068705-250MG 068705-1G
101937-312EA 553.85 USD
101937-312 101937-316
4-Chloro-7H-pyrrolo[2,3-d]pyrimidine-5-carboxylic acid ≥95%
4-Chloro-7H-pyrrolo[2,3-d]pyrimidine-5-carboxylic acid

Hazard: Irritant

Formula: C₇H₄ClN₃O₂
MW: 197.5800 g/mol
MDL Number: MFCD11518908
CAS Number: 186519-92-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR